LIF432341373 Life Chemicals Inc.

2-methoxy-5-nitrobenzoic acid

  • MDL number MFCD00527902
  • Cas Number : 40751-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart