LIF432341381 Life Chemicals Inc.

ethyl 3-aminobenzo[b]thiophene-2-carboxylate

  • MDL number MFCD00661903
  • Cas Number : 34761-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart