LIF432341382 Life Chemicals Inc.

4-(N,N-dibutylsulfamoyl)benzoic acid

  • MDL number MFCD01661020
  • Cas Number : 547-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart