LIF432341393 Life Chemicals Inc.

3-(1H-Imidazol-1-yl)propanoic acid hydrochloride

  • MDL number MFCD05884968
  • Cas Number : 96144-21-7
  • Molecular Weight : 176.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart