LIF432341403 Life Chemicals Inc.

tetrahydropyrimidin-2(1H)-imine hydrochloride

  • MDL number MFCD08704591
  • Cas Number : 26893-39-0
  • Molecular Weight : 135.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart