LIF432341406 Life Chemicals Inc.

1-(piperidin-2-yl)ethan-1-one hydrochloride

  • MDL number MFCD11506187
  • Cas Number : 106318-66-5
  • Molecular Weight : 163.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart