LIF432341407 Life Chemicals Inc.

2-methyl-6,7,8,9-tetrahydro-4H-pyrimido[1,2-a]pyrimidin-4-one

  • MDL number MFCD20689872
  • Cas Number : 879499-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart