LIF432341411 Life Chemicals Inc.

1,2,3,4,7,8,9,10-octahydro-6H-pyrimido[2,1-b]quinazolin-6-one

  • MDL number MFCD24600511
  • Cas Number : 358979-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart