LIF432341412 Life Chemicals Inc.

2-phenyl-6,7,8,9-tetrahydro-4H-pyrimido[1,2-a]pyrimidin-4-one

  • MDL number MFCD25347834
  • Cas Number : 74411-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart