LIF432341416 Life Chemicals Inc.

2-(2,5-dimethyl-1H-indol-3-yl)-N-(thiophen-2-ylmethyl)ethan-1-amine hydrochloride

  • MDL number MFCD04448790
  • Cas Number : 1052413-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart