LIF432341422 Life Chemicals Inc.

2-chloro-4-(trifluoromethyl)benzo[d]thiazole

  • MDL number MFCD08459019
  • Cas Number : 898748-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart