LIF432341428 Life Chemicals Inc.

4,6-Dimethyl-benzothiazol-2-ylamine

  • MDL number MFCD00022815
  • Cas Number : 64036-71-1
  • Molecular Weight : 178.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart