LIF432341429 Life Chemicals Inc.

1-chloro-4-phenylphthalazine

  • MDL number MFCD00524786
  • Cas Number : 10132-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart