LIF432341430 Life Chemicals Inc.

4-(aminomethyl)phthalazin-1(2H)-one hydrochloride

  • MDL number MFCD08137412
  • Cas Number : 49837-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart