LIF432341433 Life Chemicals Inc.

6,8-dimethyl-1,2,3,4-tetrahydro-1,5-naphthyridine

  • MDL number MFCD14704083
  • Cas Number : 103394-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart