LIF432341438 Life Chemicals Inc.

6-amino-5-hydroxynicotinic acid

  • MDL number MFCD21091452
  • Cas Number : 1219026-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart