LIF432341440 Life Chemicals Inc.

3-bromo-1,5-naphthyridin-4-amine

  • MDL number MFCD18806391
  • Cas Number : 91477-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart