LIF432341445 Life Chemicals Inc.

2-bromo-N,N-dimethyl-5,6,7,8-tetrahydro-1,5-naphthyridin-4-amine

  • MDL number MFCD26940451
  • Cas Number : 1706431-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart