LIF432341447 Life Chemicals Inc.

6-bromo-8-(4-methylpiperazin-1-yl)-1,2,3,4-tetrahydro-1,5-naphthyridine

  • MDL number MFCD18325131
  • Cas Number : 1263212-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart