LIF432341448 Life Chemicals Inc.

6-bromo-8-(pyrrolidin-1-yl)-1,2,3,4-tetrahydro-1,5-naphthyridine

  • MDL number MFCD28016967
  • Cas Number : 1706439-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart