LIF432341449 Life Chemicals Inc.

2-bromo-1,8-naphthyridin-3-amine

  • MDL number MFCD14704084
  • Cas Number : 1219026-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart