LIF432341450 Life Chemicals Inc.

2-bromo-1,5-naphthyridin-3-amine

  • MDL number MFCD17168221
  • Cas Number : 1219022-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart