LIF432341451 Life Chemicals Inc.

2-bromo-7-(bromomethyl)-1,8-naphthyridine

  • MDL number MFCD28165962
  • Cas Number : 1787905-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart