LIF432341452 Life Chemicals Inc.

2-bromo-7-methyl-1,8-naphthyridine

  • MDL number MFCD14705079
  • Cas Number : 1172616-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart