LIF432341453 Life Chemicals Inc.

5-amino-4-methylnicotinonitrile

  • MDL number MFCD28125843
  • Cas Number : 1706461-92-8
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart