LIF432341454 Life Chemicals Inc.

3-bromo-5-iodo-4-methylpyridine

  • MDL number MFCD17168222
  • Cas Number : 1245536-23-5
  • Molecular Weight : 297.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart