LIF432341455 Life Chemicals Inc.

8-amino-1,7-naphthyridine-5-carbonitrile

  • MDL number MFCD28125844
  • Cas Number : 1706448-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart