LIF432341458 Life Chemicals Inc.

3-bromo-5-iodo-2,4-dimethylpyridine

  • MDL number MFCD26383853
  • Cas Number : 1820612-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart