LIF432341462 Life Chemicals Inc.

(2-phenoxyethyl)amine hydrochloride

  • MDL number MFCD08582762
  • Cas Number : 17959-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart