LIF432341466 Life Chemicals Inc.

6-phenylpyridazine-3-thiol

  • MDL number MFCD09039301
  • Cas Number : 51841-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart