LIF432341467 Life Chemicals Inc.

[2-(4-fluorophenoxy)ethyl]amine hydrochloride

  • MDL number MFCD09971613
  • Cas Number : 263409-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart