LIF432341474 Life Chemicals Inc.

3,4-dihydro-2H-1,5-benzodioxepine-7-sulfonyl chloride

  • MDL number MFCD02681959
  • Cas Number : 321309-38-0
  • Molecular Weight : 248.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart