LIF432341475 Life Chemicals Inc.

[6-(4-chlorophenyl)imidazo[2,1-b][1,3]thiazol-3-yl]acetic acid

  • MDL number MFCD06810658
  • Cas Number : 68347-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart