LIF432341479 Life Chemicals Inc.

2-methoxyquinoline-4-carboxylic acid

  • MDL number MFCD01550057
  • Cas Number : 10222-62-5
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart