LIF432341481 Life Chemicals Inc.

5-iodo-2-nitrobenzoic acid

  • MDL number MFCD07783659
  • Cas Number : 35674-28-3
  • Molecular Weight : 293.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart