LIF432341484 Life Chemicals Inc.

6-bromo-1,3-dihydro-2-benzofuran-1-one

  • MDL number MFCD03428561
  • Cas Number : 19477-73-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart