LIF432341485 Life Chemicals Inc.

5-bromo-2-formylbenzoic acid

  • MDL number MFCD06739600
  • Cas Number : 4785-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart