LIF432341491 Life Chemicals Inc.

2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxamide

  • MDL number MFCD00122463
  • Cas Number : 77651-38-8
  • Molecular Weight : 182.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart