LIF432341495 Life Chemicals Inc.

4-(azepan-1-ylsulfonyl)benzoyl chloride

  • MDL number MFCD06496333
  • Cas Number : 890592-48-0
  • Molecular Weight : 301.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart