LIF432341500 Life Chemicals Inc.

8-nitroquinoline-2-carbaldehyde

  • MDL number MFCD06740734
  • Cas Number : 68311-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart