LIF432341505 Life Chemicals Inc.

2-ethoxyquinoline-3-carbaldehyde

  • MDL number MFCD04625432
  • Cas Number : 731001-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart