LIF432341522 Life Chemicals Inc.

2-(4-Methoxy-phenyl)-2-piperidin-1-yl-ethylamine dihydrochloride

  • MDL number MFCD09698502
  • Cas Number : 1093199-31-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart