LIF432341523 Life Chemicals Inc.

4-methylquinoline-2-thiol

  • MDL number MFCD00461554
  • Cas Number : 4437-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart