LIF432341524 Life Chemicals Inc.

3-chloro-1lambda6,2-benzothiazole-1,1-dione

  • MDL number MFCD00835585
  • Cas Number : 567-19-1
  • Molecular Weight : 201.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart