LIF432341526 Life Chemicals Inc.

3-Chloropyridazine trifluoromethanesulfonate

  • MDL number MFCD09881376
  • Cas Number : 1174307-32-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart