LIF432341530 Life Chemicals Inc.

4-phenyl-3-(trifluoromethyl)-1H-pyrazol-5-amine hydrochloride

  • MDL number MFCD16661089
  • Molecular Weight : 263.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-phenyl-3-(trifluoromethyl)-1H-pyrazol-5-ylamine hydrochloride
Inchi Code 1S/C10H8F3N3.ClH/c11-10(12,13)8-7(9(14)16-15-8)6-4-2-1-3-5-6;/h1-5H,(H3,14,15,16);1H
Inchi Key FIMVFCPHRHGIDS-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart