LIF432341536 Life Chemicals Inc.

{[5-(1,3-benzodioxol-5-yl)isoxazol-3-yl]methyl}amine hydrochloride

  • MDL number MFCD16660435
  • Cas Number : 1351648-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart