LIF432341538 Life Chemicals Inc.

cyclobutylhydrazine hydrochloride

  • MDL number MFCD06658415
  • Cas Number : 158001-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart