LIF432341539 Life Chemicals Inc.

1H-indol-3-ylmethanamine hydrochloride

  • MDL number MFCD19381573
  • Cas Number : 1266692-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart