LIF432341540 Life Chemicals Inc.

quinolin-2-ylmethanamine

  • MDL number MFCD02853685
  • Cas Number : 5760-20-3
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart